Protector® Evidence Drying Cabinet, Labconco
Product Details & Documents
The durable, stainless steel-lined Protector® Evidence Drying Cabinet provides a secure ventilated area to dry evidence such as clothing and bedding
Protector Evidence Drying Cabinet with Washdown features a spray nozzle that allows the interior to be washed down for easy clean up and after the cabinet has been properly disinfected. A built-in water pump aids clean up. Note that Evidence Drying Cabinet units with Washdown do not include UV lights.
Protector® Benchtop Evidence Drying Cabinets provide a compact, space-saving method of securely drying evidence. The cabinet's HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors. The key-activated ultraviolet light provides secondary disinfection of the interior between cases to prevent cross contamination, ensuring integrity of samples for DNA testing.
A variety of accessories for Protector Evidence Drying Cabinets are available, including filters, cleaning supplies, drain kits for washdown units and more. Choose from a variety of accessories for all sizes of Protector Evidence Drying Cabinets. In applicable instances, note the size of the cabinet when ordering prefilters.
The cabinet's HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors. The key-activated ultraviolet light provides secondary disinfection of the interior between cases to prevent cross contamination, ensuring integrity of samples for DNA testing. The lockable cabinet maintains the chain of custody. Full-length cabinets are available in 3', 4' and 5' widths and benchtop cabinets in 3' width. 3' cabinets have a single door and include 2" diameter locking casters. 4' and 5' cabinets have two doors and include fixed leveling feet.
Certifications: ETL
Spécifications
- Désignation:Protector Benchtop Evidence Drying Cabinet
- Taille:3'
- Electrical:230 V (Schuko)
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Hanging Clips
Informations sur le produit
- Catalog #AvailabilityPrice
- Exhaust Connection Kit
Informations sur le produit
- Catalog #AvailabilityPrice
- Paper liners, tube of 25
Informations sur le produit
- Catalog #AvailabilityPrice
- Drying Cabinet Starter Kit
Informations sur le produit
- Catalog #AvailabilityPrice
- Prefilters, Inlet, Replacement
Informations sur le produit
- Catalog #AvailabilityPrice
- Paper Liners, Tube of 100
Informations sur le produit
- Catalog #AvailabilityPrice
- Security Tags
Informations sur le produit
- Catalog #AvailabilityPrice
- Drain Kit for Washdown Units
Informations sur le produit
- Catalog #AvailabilityPrice
- Prefilters, Exhaust, Replacement
Informations sur le produit
- Catalog #AvailabilityPrice
- Cabinet Prefilter, pkg. 10
Informations sur le produit
- Catalog #AvailabilityPrice
- Paper Liners for 5' EDC, pkg. 100
Informations sur le produit
- Catalog #AvailabilityPrice
- Door Prefilter, pkg. 10
Informations sur le produit
- Catalog #AvailabilityPrice
- Floor Paper Liners for 4' EDC, pkg. 100Floor Paper Liners for 4' EDC, pkg. 10089260-186Log in pour la disponibilitéPack of 100 - $1,140.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Floor Paper Liners for 5' EDC, pkg. 25
Informations sur le produit
- Catalog #AvailabilityPrice
- Floor Paper Liners for 4' EDC, pkg. 25
Informations sur le produit
- Catalog #AvailabilityPrice
- Starter Kit for 5' EDC
Informations sur le produit
- Catalog #AvailabilityPrice
- Starter Kit for 4' EDC
Informations sur le produit
- Catalog #AvailabilityPrice
- Door Prefilter for 5' EDC, pkg. 6
Informations sur le produit
- Catalog #AvailabilityPrice
- Door Prefilter for 4' EDC, pkg. 6
Informations sur le produit
- Catalog #AvailabilityPrice
- Door Evidence Prefilter for 5' EDC, pkg. 10Door Evidence Prefilter for 5' EDC, pkg. 1089260-172Log in pour la disponibilitéPack of 10 - $123.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Door Evidence Prefilter for 4' EDC, pkg. 10Door Evidence Prefilter for 4' EDC, pkg. 1089260-170Log in pour la disponibilitéPack of 10 - $123.00