pH Electrode Solutions, WTW
pH Electrode Solutions, WTW
Pepsin Cleaning Solution, 3 x 250 ml, PEP/pH
Catalog # 89207-612
Supplier:   WTW
Supplier #:  109648
pH Electrode Solutions, WTW
Catalog # 89207-612
Supplier:  WTW
CAS Number:  
pH Electrode Solutions, WTW
Pepsin Cleaning Solution, 3 x 250 ml, PEP/pH
CAS Number:  
Price
$352.17
Log in for availability
Each (250ml)
Product Details & Documents
Solutions for operation, cleaning and storage of electrodes - simple refilling and safe storage warranted.
Electrolytes are used as refill solutions for liquid filled electrodes as well as a storage solution for all types of pH electrodes. With the pepsin cleaning solution, you can remove protein precipitation from the diaphragm of electrodes with liquid electrolyte.
Specifications
Specifications
  • Désignation:
    Pepsin Cleaning Solution, 3 x 250 ml, PEP/pH
Specifications