Objectives for Motic AE2000 Inverted Compound Microscope, Motic Instruments
Objectives for Motic AE2000 Inverted Compound Microscope, Motic Instruments
CCIS Plan Achromat PL 10X N.A. 0.25 / W.D. 16.8
Catalog # CA89168-396
Supplier:   Motic
Supplier #:  1101001703271
Objectives for Motic AE2000 Inverted Compound Microscope, Motic Instruments
Catalog # CA89168-396
Supplier:  Motic
Objectives for Motic AE2000 Inverted Compound Microscope, Motic Instruments
Objectives for Motic AE2000 Inverted Compound Microscope, Motic Instruments
CCIS Plan Achromat PL 10X N.A. 0.25 / W.D. 16.8
Price
$408.90
Log in for availability
Each
Product Details & Documents
Specifications
  • Désignation:
    CCIS Plan Achromat PL 10X N.A. 0.25 / W.D. 16.8