Objectives for Motic AE2000 Inverted Compound Microscope, Motic Instruments
CCIS Plan Achromat PL 10X N.A. 0.25 / W.D. 16.8
Catalog # CA89168-396
Supplier: Motic
Supplier #: 1101001703271
Objectives for Motic AE2000 Inverted Compound Microscope, Motic Instruments
Catalog # CA89168-396
Supplier: Motic
Jump to:
Product Details & Documents
Specifications
- Désignation:CCIS Plan Achromat PL 10X N.A. 0.25 / W.D. 16.8