ELGA Deionization Systems
Deionizer replacement cartridge, for Vision 125, filter
CA89204-278
: ELGA CA
Supplier #: VC125
ELGA’s range of wall mounted deionization systems produce from 30 to 90 liters per hour of high purity water for a wide range of laboratory and general manufacturing applications.
- Cost-effective
- Easy monitoring
- Built-in meters
- Water Quality: Type II
The deionizers are cost-effective stand-alone systems that provide low to medium volumes of primary or deionized grade water. Water quality is easy to monitor, with built in meters or colour change indicators.
- Désignation:Deionizer replacement cartridge, for Vision 125, filter