Safety Containers
Disposal can, 5 gal., dtainless dteel faucet, 5 gal.
Catalog # 89203-932
Supplier: Justrite
Supplier #: 12772
Jump to:
Product Details & Documents
Safely contain hazardous liquids and control vapors to reduce the risk of fire, protecting both personnel and facilities.
- Safely contain hazardous liquids and control vapors to reduce the risk of fire
- Helps comply with OSHA regulations and state/local fire codes
Cease-Fire® waste receptacles provide protection against trash fires.
Helps comply with OSHA regulations and state and local fire codes.
Improve efficiency with ease-of-use features which enhance pouring and filling operations.
Specifications
- Capacité:5 gal.
- Désignation:Disposal can, 5 gal., dtainless dteel faucet
- Certification:FM
Accessories
Product Information
- Catalog #AvailabilityPrice
- Extinguishing Head
Product Information
- Catalog #AvailabilityPrice
- Tubing, 3/8", Stainless Steel, EDPM Seals
Product Information
- Catalog #AvailabilityPrice
- Disconnect Kit Replacement Stainless Steel
Product Information
- Catalog #AvailabilityPrice
- Disconnect Kit Replacement Polypropylene
Product Information
- Catalog #AvailabilityPrice
- Stainless Steel Disconnect Replacement For 12018
Product Information
- Catalog #AvailabilityPrice
- Funnelspout Safety Can
Product Information
- Catalog #AvailabilityPrice
- Hose Galvanized Safety Can
Product Information
- Catalog #AvailabilityPrice
- Tubing, 3/8", Stainless Steel, VITON Seals
Product Information
- Catalog #AvailabilityPrice
- Disconnect Assembly Stainless Steel, 1/4" FittingDisconnect Assembly Stainless Steel, 1/4" Fitting14222-214Log in for availabilityEach - $1,016.71
Product Information
- Catalog #AvailabilityPrice
- Tubing, 3/8", Stainless Steel, PTFE Seals
Product Information
- Catalog #AvailabilityPrice
- Tubing, 1/4", 3/8"
Product Information
- Catalog #AvailabilityPrice
- Spill Basin Hplc Can