Safety Containers
Safety Containers
Disposal can, 5 gal., dtainless dteel faucet, 5 gal.
Catalog # 89203-932
Supplier:   Justrite
Supplier #:  12772
Safety Containers
Catalog # 89203-932
Supplier:  Justrite
Safety Containers
Safety Containers
Disposal can, 5 gal., dtainless dteel faucet, 5 gal.
Price
$1,514.05
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Safely contain hazardous liquids and control vapors to reduce the risk of fire, protecting both personnel and facilities.
  • Safely contain hazardous liquids and control vapors to reduce the risk of fire
  • Helps comply with OSHA regulations and state/local fire codes
Cease-Fire® waste receptacles provide protection against trash fires.
Helps comply with OSHA regulations and state and local fire codes.
Improve efficiency with ease-of-use features which enhance pouring and filling operations.
Specifications
  • Capacité:
    5 gal.
  • Désignation:
    Disposal can, 5 gal., dtainless dteel faucet
  • Certification:
    FM
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Extinguishing Head
    Extinguishing Head
    56610-471
    Log in for availability
    Each - $168.17
Product Information
  • Catalog #
    Availability
    Price
  • Tubing, 3/8", Stainless Steel, EDPM Seals
    Tubing, 3/8", Stainless Steel, EDPM Seals
    14222-224
    Log in for availability
    Each - $1,444.62
Product Information
  • Catalog #
    Availability
    Price
  • Disconnect Kit Replacement Stainless Steel
    Disconnect Kit Replacement Stainless Steel
    89087-386
    Log in for availability
    Each - $1,375.83
Product Information
  • Catalog #
    Availability
    Price
  • Disconnect Kit Replacement Polypropylene
    Disconnect Kit Replacement Polypropylene
    89087-388
    Log in for availability
    Each - $352.90
Product Information
  • Catalog #
    Availability
    Price
  • Stainless Steel Disconnect Replacement For 12018
    Stainless Steel Disconnect Replacement For 12018
    89178-228
    Log in for availability
    Each - $1,538.56
Product Information
  • Catalog #
    Availability
    Price
  • Funnelspout Safety Can
    Funnelspout Safety Can
    71990-050
    Log in for availability
    Each - $38.42
Product Information
  • Catalog #
    Availability
    Price
  • Hose Galvanized Safety Can
    Hose Galvanized Safety Can
    71990-054
    Log in for availability
    Each - $186.40
Product Information
  • Catalog #
    Availability
    Price
  • Tubing, 3/8", Stainless Steel, VITON Seals
    Tubing, 3/8", Stainless Steel, VITON Seals
    14222-220
    Log in for availability
    Each - $1,444.72
Product Information
  • Catalog #
    Availability
    Price
  • Disconnect Assembly Stainless Steel, 1/4" Fitting
    Disconnect Assembly Stainless Steel, 1/4" Fitting
    14222-214
    Log in for availability
    Each - $1,016.71
Product Information
  • Catalog #
    Availability
    Price
  • Tubing, 3/8", Stainless Steel, PTFE Seals
    Tubing, 3/8", Stainless Steel, PTFE Seals
    14222-216
    Log in for availability
    Each - $1,258.01
Product Information
  • Catalog #
    Availability
    Price
  • Tubing, 1/4", 3/8"
    Tubing, 1/4", 3/8"
    14222-222
    Log in for availability
    Each - $1,444.62
Product Information
  • Catalog #
    Availability
    Price
  • Spill Basin Hplc Can
    Spill Basin Hplc Can
    89212-342
    Log in for availability
    Each - $556.16