VWR® Refrigerated Circulating Baths
VWR® Refrigerated Circulating Baths offer precise temperature control, at temperature ranges from -40° to 200°C (depending on the controller).
- Offer precise temperature control
- Available in a wide range of sizes
- Three controller options
Available in a wide range of sizes, with three controller options, these refrigerated circulating baths are ideal for many applications.
The large, LCD display on the MX temperature controller is easily viewed across the lab. The Advanced Digital temperature controller offers a touch-pad, while the Advanced Programmable Temperature Controller includes a full touch-screen display, and programmability (ten 100-step programs).
The Advanced Series Controllers offer Swivel 180™ technology, which allows you to rotate the controller independently from the bath. All heated circulating baths include LidDock™ lid stowing system and DuraTop™, the precision molded phenolic bath top, which remains cooler at high temperatures and resists laboratory chemicals.
Certifications: ETL listed. CE marked. MX Controller complies with DIN 12876-1 Class I safety requirements for use with non-flammable liquids. Advanced Digital and Advanced Programmable comply with DIN 12876-1 Class III safety requirements for use with flammable liquids. cETLus compliant.
Ordering information: Refrigerated Circulators include reservoir cover; bypass tubing; male inlet and outlet adapters for 3/8, 1/4" and 3/16" (.95, .63 and .47 cm) tubing. 1/4" NPT to M16 adapters also included on 50 Hz models.
Services: Every VWR® ISO 17025 NIST Traceable Pre-calibrated Circulating Bath is calibrated and tested by an ISO 17025 accredited lab to ensure the highest standard of accuracy and traceability. Two options are available: Option 1: Standard Pre-Calibrated, Model circulating bath is calibrated and tested at both 37°C and 70°C. Option 2: Customer specified pre-calibrated circulating bath provides a choice of 3 different temperature points based on your research requirements. Simply order the stated catalog number and you will receive a protective box containing a work order form allowing you to specify your own calibration temperature points. After completing the work order, you can mail the box using a pre-paid return shipping label or you can scan and email the information to a specified E-mail address. The Avantor calibration team will calibrate the circulating bath based on your instruction.
VWRCATALYST™ can provide an array of services, such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWR[email protected] for further details.
- Plage de température:–40...+200 °C
- Désignation:15L Refrigerated Circulator with Advanced Programmable Controller, -40°C
Accessories
- DescriptionSizeFor Use With
- 13270-792PolyScience polytherm S2503.8 l (1 gal.)VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath SystemsEach (1US Gal) - $3,876.06
- DescriptionSizeFor Use With
- 13270-794PolyScience polytherm S1503.8 l (1 gal.)VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath SystemsEach (1US Gal) - $2,893.65
- DescriptionSizeFor Use With
- 13272-034PolyScience polycool HC -50, Freezing Point –56 °C3.8 l (1 gal.)VWR Refrigerated Circulating BathsEach (1US Gal) - $424.81
- DescriptionFor Use WithDetails
- 71002-500PolyScience polyclean CLARIFIERVWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systems8 oz bottle of polyclean CLARIFIER; concentrated; treats approximately 200 gallons (757 liters)Each (8oz) - $83.31
- DescriptionTemperature Range
- 71002-504Ethylene glycol, 3.8 l (1 gal.)–25...+100 °C (–13...+212 °F), when mixed 50/50 with distilled waterEach (1US Gal) - $253.34
- DescriptionFor Use WithDetails
- 71002-502PolyScience polyclean CLARIFIERVWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systemscase of 12 (8 oz bottles); polyclean CLARIFIER; concentrated; treats appromixately 200 gallons (757 liters)Case of 12 (1/Itm/Bx) - $952.76
- DescriptionTemperature Range
- 71002-506Propylene glycol, 3.8 l (1 gal.)–20...+100 °C (–4...+212 °F), when mixed 50/50 with distilled waterEach (1US Gal) - $457.69
- DescriptionFor Use WithDetails
- 89201-0066" External Pt100 Probe with 7.6 m (25') length cableVWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersFor Monitoring the Temperature of an External ApplicationEach - $1,029.25
- DescriptionFor Use WithDetails
- 89201-0046" External Pt100 Probe with 3 m (10') length cableVWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersFor Monitoring the Temperature of an External ApplicationEach - $743.10
- DescriptionFor Use WithDetails
- 89201-0086" External Pt100 Probe with 15 m (50') length cableVWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersFor Monitoring the Temperature of an External ApplicationEach - $1,552.33
- DescriptionFor Use WithDetails
- 89201-010RS232 CableVWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersCable Length: 9'Each - $101.37
- DescriptionFor Use WithDetails
- 89201-0561/4" ID Buna N TubingVWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systems–40...120 °C, Order in Continuous Lengths Of 3' (1 m)Each - $33.01
- DescriptionFor Use WithDetails
- 89201-012Ethernet CableVWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersCable Length: 9'Each - $88.59
- DescriptionFor Use WithDetails
- 89201-062Insulation for 1/4" TubingVWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath SystemsOrder in Continuous Lengths of 2 Yards (6 ft.)Each - $143.30
- DescriptionFor Use WithDetails
- 89201-076Tube ClampsVWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath SystemsFor 1/4 and 3/8" ID Tubing, Stainless SteelEach - $16.46