Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Sample Vials with Attached Closures, DWK Life Sciences

Sample Vials with Attached Closures, DWK Life Sciences

40.00 ml, 28×95 mm, Clear, Oui, 24-400, Black, 72/case
Catalog # 89201-744
Supplier #:  60811B-10
Sample Vials with Attached Closures, DWK Life Sciences
Catalog # 89201-744
Sample Vials with Attached Closures, DWK Life Sciences
Sample Vials with Attached Closures, DWK Life Sciences

40.00 ml, 28×95 mm, Clear, Oui, 24-400, Black, 72/case

Price
$455.00
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 72
Product Details & Documents

Screw thread sample vials with attached closures manufactured from clear 33 expansion borosilicate glass.

  • Closed top black phenolic closures attached
  • PTFE-faced white rubber liner or white rubber liner

Packaging: Vials are packaged in corrugated trays with partitions and small case quantities.

Spécifications
  • Avec bouchon:
    Oui
  • Taille du bouchon:
    24-400
  • Capacité:
    40.00 ml
  • Color:
    Clear
  • Packaging:
    72/case
  • Diameter ×H:
    28×95 mm
  • Cap Color:
    Black
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.