BD Eclipse™ Needles and Syringes, BD
BD Eclipse™ Needles and Syringes, BD
BD Luer-Lok Syringe with Detachable Needle, 1 ml, 25, 5/8"
Catalog # BD305780
Supplier #:  305780
BD Eclipse™ Needles and Syringes, BD
Catalog # BD305780
Supplier:  
BD Eclipse™ Needles and Syringes, BD
BD Eclipse™ Needles and Syringes, BD
BD Luer-Lok Syringe with Detachable Needle, 1 ml, 25, 5/8"
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
BD ECLIPSE COMBO 1ML 25GX5/8IN PK50 Catalog #CABD305780
Product CABD305780 is a replacement for BD305780 and has the same packaging and specifications, but may be a different quantity.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
BD Eclipse™ Needles are designed with pivoting needle technology for single-handed activation
For use with BD Luer-Lok™ Syringes.
No hard surface is required with this needle-based, safety-engineered injection device.
Spécifications
  • Volume:
    1 ml
  • Désignation:
    BD Luer-Lok Syringe with Detachable Needle
  • Needle Length:
    5/8"
  • Jauge:
    25