Casters for Modular Workstations
Roll and set casters, set of 4, 1100 lbs.
Catalog # 89202-114
Jump to:
Discontinued
Product Details & Documents
Make any Sovella lab bench a mobile lab station by adding casters! Work is not always confined to one location.
- Contain foot activated total lock brake
- Easy swiveling and smooth rolling
- Durable zinc plated finish
Ordering information: Casters sold in sets of four. Installation is required. Assembly videos available.
Spécifications
- Désignation:Roll and set casters, set of 4
- Load Capacity:1100 lbs.