Casters for Modular Workstations
Lab caster set of 4 (2 locking, 2 non-locking), 660 lbs.
89202-110
: Treston
: 834734-00
Make any Sovella lab bench a mobile lab station by adding casters! Work is not always confined to one location.
- Contain foot activated total lock brake
- Easy swiveling and smooth rolling
- Durable zinc plated finish
: Casters sold in sets of four. Installation is required. Assembly videos available.
- Désignation:Lab caster set of 4 (2 locking, 2 non-locking)
- Load Capacity:660 lbs.