Mod-Rack™ Pegboard, Interdyne
24", 24", 20
Catalog # VF9-V2424
Supplier: Interdyne
Supplier #: V2424
Jump to:
Product Details & Documents
Stainless steel pegboard features interchangeable white polypropylene pegs, integral drip trough, and clear plastic drain tube
Completely autoclavable and stain-resistant. Wall bracket included.
Ordering information: Accessories including a funnel rack, drain basket, screen insert, and pipet rack are also available; contact your Avantor sales representative for more information.
Specifications
- Largeur:24"
- Hauteur:24"
- No. of Pegs:20
Accessories
Product Information
- Catalog #AvailabilityPrice
- Screen Insert for Drain Trough, Stainless Steel
Product Information
- Catalog #AvailabilityPrice
- Screen Insert for Drain Trough, Stainless Steel
Product Information
- Catalog #AvailabilityPrice
- Drain Basket for Small Glassware Items
Product Information
- Catalog #AvailabilityPrice
- Petite C-Fold Paper Towel Dispenser
Product Information
- Catalog #AvailabilityPrice
- Vertical Liquid Soap Dispenser