Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Cleanroom Labels | Avantor

Cleanroom Labels

Direct thermal cleanroom labels, 76×51 mm (3×2"), 1000 labels/roll
Catalog # TWTX532D
Supplier:   Texwipe
Fournisseur #:  TX532D
Cleanroom Labels
Catalog # TWTX532D
Supplier:  Texwipe
Cleanroom Labels
Cleanroom Labels

Direct thermal cleanroom labels, 76×51 mm (3×2"), 1000 labels/roll

Price
399,50 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Roll of 1,000 Case of 4000
Product Details & Documents

Designed specifically for cleanroom label printing applications requiring direct heat printing.

  • Permanent adhesive

A polypropylene material. Available with two different adhesive levels to provide excellent adhesion without leaving residue.

Spécifications
  • Désignation:
    Direct thermal cleanroom labels
  • Dimensions:
    76×51 mm (3×2")
  • Packaging:
    1000 labels/roll
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.