KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
25 ml, Pear-shaped flask, threaded neck, O.D×H: 80×39 mm
Catalog # KT747540-2520
Supplier: DWK Life Sciences (KIMBLE)
Fournisseur #: 747540-2520
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
Catalog # KT747540-2520
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
These flasks have short necks for easy retrieval of residues or reaction products.
- Accessory for Microscale MICROFLEX® threaded kits
- Thread size: 20-400
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Spécifications
- Capacité:25 ml
- Désignation:Pear-shaped flask, threaded neck, O.D×H: 80×39 mm