KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
15 ml, Pear-shaped flask, threaded neck, O.D×H: 30×55 mm
  KT747540-1520
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
  KT747540-1520
 :  
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
15 ml, Pear-shaped flask, threaded neck, O.D×H: 30×55 mm
These flasks have short necks for easy retrieval of residues or reaction products.
  • Accessory for Microscale MICROFLEX® threaded kits
  • Thread size: 20-400
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
  • Capacité:
    15 ml
  • Désignation:
    Pear-shaped flask, threaded neck, O.D×H: 30×55 mm