KONTES® Round-Bottom Boiling Flasks, Threaded, Kimble Chase
KONTES® Round-Bottom Boiling Flasks, Threaded, Kimble Chase
5 ml, 27×40 mm
Catalog # KT747510-0520
Supplier #:  747510-0520
KONTES® Round-Bottom Boiling Flasks, Threaded, Kimble Chase
Catalog # KT747510-0520
KONTES® Round-Bottom Boiling Flasks, Threaded, Kimble Chase
KONTES® Round-Bottom Boiling Flasks, Threaded, Kimble Chase
5 ml, 27×40 mm
Price
$36.86
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
All flasks have a short neck and 20-400 GPI thread size.
Specifications
  • Capacité:
    5 ml
  • Diameter ×H:
    27×40 mm