Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
KIMBLE® Separatory Funnels, Squibb Type, DWK Life Sciences | Avantor

KIMBLE® Separatory Funnels, Squibb Type, DWK Life Sciences

Seporatory funnel with 16 stopper, 60 ml, 223 mm
Catalog # KT636030-0060
Fournisseur #:  636030-0060
KIMBLE® Separatory Funnels, Squibb Type, DWK Life Sciences
Catalog # KT636030-0060
KIMBLE® Separatory Funnels, Squibb Type, DWK Life Sciences
KIMBLE® Separatory Funnels, Squibb Type, DWK Life Sciences

Seporatory funnel with 16 stopper, 60 ml, 223 mm

Price
$321.23
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents

KONTES® Squibb separatory funnels with PTFE stopcock and standard taper joint.

  • Plain stem, but may be used with PTFE BEVEL-SEAL™ inlet adapter (Note stem diameter for a proper fit when using this adapter)
  • With an 1:5 taper PTFE stopcock plug
  • Manufactured from 33 expansion, low extractable borosilicate glass

KT636031-1000 is supplied without a stopper. KIMBLE® separatory funnels, squibb type (KT636030 series) is supplied with a glass stopper.

Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.

Documents

Spécifications
  • Capacité:
    60 ml
  • Désignation:
    Seporatory funnel with 16 stopper
  • Hauteur:
    223 mm