Accessories
Polypropylene Packing Reservoirs, 700 mL, for 2.5 cm I.D. Columns
Catalog # KT420405-0700
Supplier: DWK Life Sciences (KIMBLE)
Fournisseur #: 420405-0700
Jump to:
Product Details & Documents
Spécifications
- Désignation:Polypropylene Packing Reservoirs, 700 mL, for 2.5 cm I.D. Columns