ContactUsLinkComponent
Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
  CAEX0995-1
undefined
Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
  CAEX0995-1
 :  EX0995-1
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Extran® MN 01 Detergent Phosphate-free
  • Size:
    1.8 kg
  • Storage temperature:
    RT
  • Cat. no.:
    CAEX0995-1
  • Packaging:
    Plastic bottle
  • Supplier No.:
    EX0995-1
When diluted with water, concentrates can clean by themselves without leaving residues
Extran MN 01 Powdered Detergent is a multi-purpose detergent for heavy-duty laboratory cleaning applications. This mixture of anionic surfactants and auxiliary agents can be used on glass, plastic, rubber, metal, and quartz. Phosphate-free. Items to be cleaned should be immersed in a cleaning solution and thoroughly brushed or wiped. Dose 2 to 5 g/L. pH of 0.2% solution is 10.9.
For routine laboratory applications, no scrubbing is necessary once items are removed from the cleaning bath. For best results, items to be cleaned should be soaked for 2 to 24 hours, and rinsed with distilled water immediately after removal from cleaning bath.
Frequently Bought Together