Supelco® pH Buffer Solutions, MilliporeSigma
Supelco® pH Buffer Solutions, MilliporeSigma
±0.02, 3.00
  CABX1653-1
Supelco® pH Buffer Solutions, MilliporeSigma
  CABX1653-1
 :  
 :  
Supelco® pH Buffer Solutions, MilliporeSigma
Supelco® pH Buffer Solutions, MilliporeSigma
±0.02, 3.00
 :  
BDH BUFFER REF STD PH3 COLORLESS 500ML  BDH5014-500ML
Product BDH5014-500ML is similar to CABX1653-1 but manufacturer and specifications may vary.
PH3 CALIB BUFFER 1 PINT 1PT  RC1495-16
Product RC1495-16 is similar to CABX1653-1 but manufacturer and specifications may vary.
Certified reference material, traceable to NIST Standard.
  • Ready-to-use solution available in a sealed bottle with convenient packaging to protect against contamination
Available as colorless or color-coded solutions.
  • pH:
    3.00
  • pH Accuracy:
    ±0.02