BD Eclipse™ Needles and Syringes, BD
BD Luer-Lok Syringe with Detachable Needle, 5 ml, 22, 11/2"
  CABD305785
BD Eclipse™ Needles and Syringes, BD
  CABD305785
 :  
BD Eclipse™ Needles and Syringes, BD
BD Eclipse™ Needles and Syringes, BD
BD Luer-Lok Syringe with Detachable Needle, 5 ml, 22, 11/2"
BD Eclipse™ Needles are designed with pivoting needle technology for single-handed activation
For use with BD Luer-Lok™ Syringes.
No hard surface is required with this needle-based, safety-engineered injection device.
  • Volume:
    5 ml
  • Désignation:
    BD Luer-Lok Syringe with Detachable Needle
  • Needle Length:
    11/2"
  • Jauge:
    22