BD Eclipse™ Needles and Syringes, BD
Product Details & Documents
BD Eclipse™ Needles are designed with pivoting needle technology for single-handed activation
For use with BD Luer-Lok™ Syringes.
No hard surface is required with this needle-based, safety-engineered injection device.
Spécifications
- Volume:3 ml
- Désignation:BD Luer-Lok Syringe with Detachable Needle
- Needle Length:5/8"
- Jauge:25