BD Eclipse™ Needles and Syringes, BD
BD Eclipse™ Needles and Syringes, BD
BD Luer-Lok Syringe with Detachable Needle, 1 ml, 25, 5/8"
Catalog # CABD305780
Supplier:   BD
Supplier #:  305780
BD Eclipse™ Needles and Syringes, BD
Catalog # CABD305780
Supplier:  BD
BD Eclipse™ Needles and Syringes, BD
BD Eclipse™ Needles and Syringes, BD
BD Luer-Lok Syringe with Detachable Needle, 1 ml, 25, 5/8"
Price
$79.50
Log in pour la disponibilité
Pack of 50 Case of 300
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
BD Eclipse™ Needles are designed with pivoting needle technology for single-handed activation
For use with BD Luer-Lok™ Syringes.
No hard surface is required with this needle-based, safety-engineered injection device.
Spécifications
  • Volume:
    1 ml
  • Désignation:
    BD Luer-Lok Syringe with Detachable Needle
  • Needle Length:
    5/8"
  • Jauge:
    25