D(-)-Ribose 98%
D(-)-Ribose 98%
Bottle, 250 g
Catalog # CAAAA17894-30
Supplier #:  A17894-30
D(-)-Ribose 98%
Catalog # CAAAA17894-30
CAS Number:  50-69-1
D(-)-Ribose 98%
D(-)-Ribose 98%
Bottle, 250 g
CAS Number:  50-69-1
Price
$613.49
Log in for availability
Each (250g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Purity: 98% CAS No.: 50-69-1 Molecular Formula: C5H10O5 Formula Weight: 150.13 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 82-86° Beilstein Reference: 1723081 Merck Reference: 14,8204 Optical Rotation: -20° (c=10 in water, 28h) MDL No.: MFCD00135453
  • CAS Number:50-69-1
  • Formula:C₅H₁₀O₅
  • MDL Number:MFCD00135453
  • Molecular Weight:150.13 g/mol
  • Merck Index:12,08371
  • Boiling Pt:331 °C (1013 hPa)
  • Melting Pt:90 - 95 °C
  • Melting Pt:82…86 °C
  • Density:0,8 g/cm³ (20 °C)
  • Storage Temperature:Température ambiante
  • EINECS:200-059-4
Specifications
Specifications
  • Type de conditionnement:
    Bottle
  • Taille:
    250 g