Ceramic Rods, Round, Solid, AL23, Thermo Scientific
1000 mm, 10 mm
  CAAA32937-EO
Ceramic Rods, Round, Solid, AL23, Thermo Scientific
  CAAA32937-EO
 :  
Ceramic Rods, Round, Solid, AL23, Thermo Scientific
1000 mm, 10 mm
Pure Al2O3 Aluminium oxide, AL23
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Solid aluminum rod.
  • Ø:
    10 mm
  • Longueur:
    1000 mm