Ceramic Rods, Round, Solid, AL23, Thermo Scientific
Product Details & Documents
Pure Al2O3 Aluminium oxide, AL23
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Solid aluminum rod.
Spécifications
- Ø:8 mm
- Longueur:200 mm
- Cat. No.:CAAA32935-DR