Ceramic Rods, Round, Solid, AL23, Thermo Scientific
200 mm, 1.2 mm
  CAAA32924-DR
Ceramic Rods, Round, Solid, AL23, Thermo Scientific
  CAAA32924-DR
 :  
Ceramic Rods, Round, Solid, AL23, Thermo Scientific
200 mm, 1.2 mm
Pure Al2O3 Aluminium oxide, AL23
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Solid aluminum rod.
  • Ø:
    1.2 mm
  • Longueur:
    200 mm