Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Electrode Fill Solutions

Electrode Fill Solutions

1M KCl, Bottle, 25 ml, —
Catalog # CA97001-716
Supplier:   Mettler Toledo
Supplier #:  51343181
Electrode Fill Solutions
Catalog # CA97001-716
Supplier:  Mettler Toledo
CAS Number:  
Electrode Fill Solutions
Electrode Fill Solutions

1M KCl, Bottle, 25 ml, —

CAS Number:  
Price
$34.50
Log in for availability
Each (25ml)
Product Details & Documents

FRISCOLYT-B™ (84.4% glycerol, 11% KCl, and 4.6% H₂O) is for low temperature samples.

 : Supplied in 25 ml (0.9 oz.) bottles.

Documents

Specifications

Specifications

  • Type de conditionnement:
    Bottle
  • Désignation:
    1M KCl
  • Ion:
  • Volume:
    25 ml

Specifications