BBL™ Standard Methods Agar, BD Life Sciences
BBL™ Standard Methods Agar, Deeps, 20 mL
Supplier: BD
: 221030
Jump to:
Product Details & Documents
Standard Methods Agar is used for obtaining microbial plate counts from milk and dairy products, foods, water, and other materials of sanitary importance.
- Enzymatic digest of casein provides nitrogen and amino acids
- Yeast extract supplies B-complex vitamins
- Dextrose is an energy source
- Agar is the solidifying agent
Standard Methods Agar is made according to the American Public Health Association (APHA) formulation.
Certificates
Recherche par numéro de lotSpécifications
Spécifications
- Désignation:BBL™ Standard Methods Agar, Deeps
- Taille:20 mL