BBL™ Standard Methods Agar, BD Life Sciences
BBL™ Standard Methods Agar, Deeps, 20 mL
Catalog # CA90001-200
Supplier:   BD
 :  221030
BBL™ Standard Methods Agar, BD Life Sciences
Catalog # CA90001-200
Supplier:  BD
CAS Number:  
BBL™ Standard Methods Agar, BD Life Sciences
BBL™ Standard Methods Agar, Deeps, 20 mL
CAS Number:  
Price
$57.60
Log in pour la disponibilité
Pack of 10
Product Details & Documents
Standard Methods Agar is used for obtaining microbial plate counts from milk and dairy products, foods, water, and other materials of sanitary importance.
  • Enzymatic digest of casein provides nitrogen and amino acids
  • Yeast extract supplies B-complex vitamins
  • Dextrose is an energy source
  • Agar is the solidifying agent
Standard Methods Agar is made according to the American Public Health Association (APHA) formulation.
Spécifications
Spécifications
  • Désignation:
    BBL™ Standard Methods Agar, Deeps
  • Taille:
    20 mL
Spécifications