R2A Agar is used for enumerating heterotrophic organisms in treated potable water.
- Dehydrated appearance: Light beige, flree-flowing
- Prepared appearance: Light amber, slightly opalescent
- Improves recovery of chlorine-tolerant bacteria
A low-nutrient medium, R2A Agar is used in combination with a lower incubation temperature and longer incubation time to stimulate the growth of stressed and chlorine-tolerant bacteria.
- Désignation:R2A Agar
- Taille:100 g