Difco™ Select APS™ Super Broth Base, BD Diagnostics
Difco™ Select APS™ Super Broth Base, BD Diagnostics
Difco™ Select APS™ Super Broth Base, 500 g
Catalog # CA90000-676
Supplier:   BD
Supplier #:  212485
Difco™ Select APS™ Super Broth Base, BD Diagnostics
Catalog # CA90000-676
Supplier:  BD
CAS Number:  
Difco™ Select APS™ Super Broth Base, BD Diagnostics
Difco™ Select APS™ Super Broth Base, 500 g
CAS Number:  
Price
$240.00
Log in for availability
Each (500g)
Product Details & Documents
Select APS™ Super Broth is a molecular genetics medium used to grow Escherichia coli to a high cell density.
  • Molecular genetics medium
  • Alternative to classical animal-based media
  • Allows maintenance and propogation of E. coli
  • Soy hydrolysate provides nitrogen and carbon
  • Glycerol is added as a carbon and energy source
Select APS™ media are manufactured from animal-free ingredients in order to minimize the risk of bovine spongiform encephalopathy in culture media containing animal, and especially bovine, materials.
Specifications
Specifications
  • Désignation:
    Difco™ Select APS™ Super Broth Base
  • Taille:
    500 g