Difco™ Select APS™ Super Broth Base, BD Diagnostics
Difco™ Select APS™ Super Broth Base, 500 g
Supplier: BD
Supplier #: 212485
Jump to:
Product Details & Documents
Select APS™ Super Broth is a molecular genetics medium used to grow Escherichia coli to a high cell density.
- Molecular genetics medium
- Alternative to classical animal-based media
- Allows maintenance and propogation of E. coli
- Soy hydrolysate provides nitrogen and carbon
- Glycerol is added as a carbon and energy source
Select APS™ media are manufactured from animal-free ingredients in order to minimize the risk of bovine spongiform encephalopathy in culture media containing animal, and especially bovine, materials.
Specifications
Specifications
- Désignation:Difco™ Select APS™ Super Broth Base
- Taille:500 g