Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
BBL™ Middlebrook OADC Enrichment, BD Life Sciences | Avantor

BBL™ Middlebrook OADC Enrichment, BD Life Sciences

Middlebrook OADC Enrichment, 500 mL
Catalog # CA90000-614
Supplier:   BD
Fournisseur #:  212351
BBL™ Middlebrook OADC Enrichment, BD Life Sciences
Catalog # CA90000-614
Supplier:  BD
CAS Number:  
BBL™ Middlebrook OADC Enrichment, BD Life Sciences
BBL™ Middlebrook OADC Enrichment, BD Life Sciences

Middlebrook OADC Enrichment, 500 mL

CAS Number:  
Price
$781.00
Log in pour la disponibilité
Each (500ml)
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Middlebrook OADC Enrichment is a supplement recommended for isolation and cultivation of Mycobacteria.

  • Enhances recovery on primary isolation
  • Dextrose is an energy source
  • Catalase destroys toxic peroxides
  • Promotes mycobacterial growth

Supplementation of the agar base is required in order to obtain mycobacterial growth. In the enriched medium, sodium chloride maintains osmotic equilibrium; oleic acid, as well as other long chain fatty acids, can be utilized by tubercle bacilli and plays an important role in the metabolism of mycobacteria; the primary effect of albumin is that of protection of the tubercle bacilli against toxic agents and, therefore, it enhances their recovery on primary isolation; dextrose is an energy source; and catalase destroys toxic peroxides that may be present in the medium.

Documents

Spécifications

Spécifications

  • Désignation:
    Middlebrook OADC Enrichment
  • Taille:
    500 mL

Spécifications