BBL™ Mannitol Salt Agar, BD
BBL™ Mannitol Salt Agar, BD
Mannitol Salt Agar, 5 lb
Catalog # CA90000-184
BBL™ Mannitol Salt Agar, BD
Catalog # CA90000-184
Supplier:  
CAS Number:  
BBL™ Mannitol Salt Agar, BD
BBL™ Mannitol Salt Agar, BD
Mannitol Salt Agar, 5 lb
CAS Number:  
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
MANNITOL SALT AGAR 25LB Catalog #CA90004-534
Product CA90004-534 is a replacement for CA90000-184 and has the same packaging and specifications, but may be a different quantity.
MANNITOL SALT AGAR 500GM Catalog #CA90000-182
Product CA90000-182 is a replacement for CA90000-184 and has the same packaging and specifications, but may be a different quantity.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mannitol Salt Agar is a selective and differential medium for the detection and enumeration of staphylococci from clinical and nonclinical specimens.
  • Beef extract and peptones provide essential growth factors
  • 7.5% concentration of sodium chloride inhibits bacterial growth
  • Mannitol fermentation aids in differentiation of staphylococcal species
Difference in colony growth colors also acts as an identifying feature.
Spécifications
Spécifications
  • Désignation:
    Mannitol Salt Agar
  • Taille:
    5 lb