Embedding Rings, Simport Scientific | Avantor
Embedding Rings, Simport Scientific
Yellow
Catalog # CA73960-006
Supplier:   Simport Scientific
Fournisseur #:  M460-5
Embedding Rings, Simport Scientific
Catalog # CA73960-006
Supplier:  Simport Scientific
Embedding Rings, Simport Scientific
Embedding Rings, Simport Scientific
Yellow
Price
$302.29
Log in pour la disponibilité
Case of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made of high-impact polystyrene, these embedding rings are suitable for holding and identifying tissue sample blocks
The rings fit well in microtome chuck adapters. The etched writing surface on the ring is marked with an identification number and placed on top of the sample block. Additional paraffin is poured into the base mold to cement the ring onto the tissue block. The embedding ring securely holds the tissue sample in the microtome chuck adapter for sectioning and then identifies the sample while in storage.
Ordering information: Each case contains four dispenser boxes of 250 rings.
Spécifications
  • Color:
    Yellow