StoreBox™ Storage Boxes, Simport
StoreBox™ Storage Boxes, Simport
StoreBox™ for 3 - 5 ml Tubes, Pink, PS, 51/4×51/4×33/4"
  CA60872-876
Supplier #:  T514-581P
StoreBox™ Storage Boxes, Simport
  CA60872-876
StoreBox™ Storage Boxes, Simport
StoreBox™ Storage Boxes, Simport
StoreBox™ for 3 - 5 ml Tubes, Pink, PS, 51/4×51/4×33/4"
$166.60
Pack of 5 Case of 10
These economical storage boxes are designed to be used at temperatures between –90 and 80 °C. Different models are available to accommodate either 25, 81, or 100 sample tubes.
  • Cover has numbered squares for easy sample identification
  • Two corners of cover and base are cut to prevent misalignment
  • Writing surface provided to identify base and/or cover
  • Vials readily visible through transparent cover
  • Air vents minimize condensation
  • Four pastel colors available for better color coding
A transparent cover allows the user to see the contents of the box. The cover is keyed to the base to prevent misalignment. Printed with a series of squares (numbered), the surface accepts writing with markers, facilitating inventory control.
A unique color coding system uses colored plastic grids to separate the cover from the base on the 25- and 81- place boxes. Those made to accept 100 tubes have a colored base instead of a grid. Vial removal is facilitated by an innovative vial picker supplied with each storage box. Not autoclavable.
  • Color:
    Pink
  • Désignation:
    StoreBox™ for 3 - 5 ml Tubes
  • Dimensions:
    51/4×51/4×33/4"
  • Matière:
    PS