Prefilter for Masks, 3M™
Prefilter for Masks, 3M™
P100 filter "nuisance odors generated from chlorine, sulfur dioxide and hydrogen fluorid, solid and liquid particles including oil" for use with 3M™ 5000, 6000, and 7000 series respirators, Provides relief from nuisance odors generated from chlorine, sulfur dioxide and hydrogen fluoride, 99.97% efficient at removing solid and liquid particles including those containing oil, use with 3M™ 5000, 6000, and 7000 series respirators
Catalog # CA56621-938
Supplier:   3M
Supplier #:  7000002048
Prefilter for Masks, 3M™
Catalog # CA56621-938
Supplier:  3M
Prefilter for Masks, 3M™
P100 filter "nuisance odors generated from chlorine, sulfur dioxide and hydrogen fluorid, solid and liquid particles including oil" for use with 3M™ 5000, 6000, and 7000 series respirators, Provides relief from nuisance odors generated from chlorine, sulfur dioxide and hydrogen fluoride, 99.97% efficient at removing solid and liquid particles including those containing oil, use with 3M™ 5000, 6000, and 7000 series respirators
Price
18,20 $
Log in for availability
Pack of 2 Case of 100
Product Details & Documents
For use with 5000-series respirators.
  • NIOSH 42 CFR 84 approved
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Specifications
  • Désignation:
    P100 filter "nuisance odors generated from chlorine, sulfur dioxide and hydrogen fluorid, solid and liquid particles including oil" for use with 3M™ 5000, 6000, and 7000 series respirators
  • Approved For Use Against:
    Provides relief from nuisance odors generated from chlorine, sulfur dioxide and hydrogen fluoride, 99.97% efficient at removing solid and liquid particles including those containing oil, use with 3M™ 5000, 6000, and 7000 series respirators