MICROSETTE™ I Biopsy Processing and Embedding Cassettes, Simport Scientific
Product Details & Documents
These innovative disposable plastic cassettes eliminate the need for biopsy pads
Cover and base feature over 2000 square openings, each with a diameter of 0.26mm. These mesh holes allow for maximum fluid exchange. The high-density acetal polymer is resistant to the chemical action of histological solvents. The integral one-piece lids can be opened and closed multiple times. The anterior writing area is set at a 45° angle. Compatible with most cassette printers.
: Packaged in dispenser boxes of 150 cassettes with covers assembled, four boxes per case.
Spécifications
- Color:Lilac