MICROSETTE™ I Biopsy Processing and Embedding Cassettes, Simport Scientific
MICROSETTE™ I Biopsy Processing and Embedding Cassettes, Simport Scientific
Yellow
Catalog # CA48648-315
Supplier:   Simport Scientific
Supplier #:  M502-5
MICROSETTE™ I Biopsy Processing and Embedding Cassettes, Simport Scientific
Catalog # CA48648-315
Supplier:  Simport Scientific
MICROSETTE™ I Biopsy Processing and Embedding Cassettes, Simport Scientific
MICROSETTE™ I Biopsy Processing and Embedding Cassettes, Simport Scientific
Yellow
Price
$251.31
Log in for availability
Pack of 250 Case of 1000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These innovative disposable plastic cassettes eliminate the need for biopsy pads
Cover and base feature over 2000 square openings, each with a diameter of 0.26mm. These mesh holes allow for maximum fluid exchange. The high-density acetal polymer is resistant to the chemical action of histological solvents. The integral one-piece lids can be opened and closed multiple times. The anterior writing area is set at a 45° angle. Compatible with most cassette printers.
 : Packaged in dispenser boxes of 150 cassettes with covers assembled, four boxes per case.
Specifications
  • Color:
    Yellow