VWR® Micro Cover Glasses, Rectangular
Cover glasses, rectangular, 60×24 mm, #1.5
Catalog # CA48393-252-1
VWR® Micro Cover Glasses, Rectangular
Catalog # CA48393-252-1
Supplier:  
VWR® Micro Cover Glasses, Rectangular
VWR® Micro Cover Glasses, Rectangular
Cover glasses, rectangular, 60×24 mm, #1.5
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
Product Details & Documents
These micro cover glasses are made from the best quality glass, and each set of numbered slides is uniform in size and thickness.
  • Quality glass construction
  • Uniform sizes
  • Two-slide thickness options available
The No. 1 slides are 0.13 to 0.17 mm in thickness. The No. 1¹/₂ slides are 0.16 to 0.19 mm in thickness.
 : Slides are packed 1 oz. per box.
Spécifications
  • Numbering:
    #1.5
  • Désignation:
    Cover glasses, rectangular
  • L×l:
    60×24 mm