Base Molds, Triangle Biomedical
Base Molds, Triangle Biomedical
Plastic Base Molds, 7 x 7 x 6 mm
Catalog # CA27900-076
Base Molds, Triangle Biomedical
Catalog # CA27900-076
Supplier:  
Base Molds, Triangle Biomedical
Base Molds, Triangle Biomedical
Plastic Base Molds, 7 x 7 x 6 mm
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Reusable Base Mold with winged edges come in stainless steel and plastic
Base molds are disposable and chemical resistant.
Spécifications
  • Désignation:
    Plastic Base Molds
  • L×W×D:
    7 x 7 x 6 mm