Nunc® CryoColor Coder Inserts, Thermo Scientific
Assorted
Supplier: Thermo Fisher Scientific
Supplier #: 375930
Nunc® CryoColor Coder Inserts, Thermo Scientific
Catalog # CA24270-120A
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
For comprehensive sample identification, fits inside CryoTube® vial cap.
- Made of polystyrene
- Available in 10 different colors
Ordering information: Assortment 24270-120 includes either 50 of each color (for a pack) or 200 of each color (for a case).
Spécifications
- Color:Assorted