Nunc® CryoColor Coder Inserts, Thermo Scientific
Nunc® CryoColor Coder Inserts, Thermo Scientific
Assorted
Catalog # CA24270-120A
Supplier #:  375930
Nunc® CryoColor Coder Inserts, Thermo Scientific
Catalog # CA24270-120A
Nunc® CryoColor Coder Inserts, Thermo Scientific
Nunc® CryoColor Coder Inserts, Thermo Scientific
Assorted
Price
$140.35
Log in pour la disponibilité
Pack of 500 Case of 2000
Product Details & Documents
For comprehensive sample identification, fits inside CryoTube® vial cap.
  • Made of polystyrene
  • Available in 10 different colors
Ordering information: Assortment 24270-120 includes either 50 of each color (for a pack) or 200 of each color (for a case).
Spécifications
  • Color:
    Assorted