Nunc® CryoColor Coder Inserts, Thermo Scientific
Grey
: 375906
For comprehensive sample identification, fits inside CryoTube® vial cap.
- Made of polystyrene
- Available in 10 different colors
: Assortment 24270-120 includes either 50 of each color (for a pack) or 200 of each color (for a case).
- Color:Grey