VWR® Corks | Avantor
VWR® Corks
Catalog # CA23420-560
VWR® Corks
Catalog # CA23420-560
Supplier:  
VWR® Corks
VWR® Corks
Discontinued
Product Details & Documents
XXXX Corks function as a closure for bottles, vials, laboratory and other openings.
  • Regular length taper
  • Assorted sizes
The XXXX premium grade corks have fewer lenticels (crevices) than standard grade
Selected grade of fine textured corks. Sizes 22 through 30 have cemented construction.
This means they are better suited for use as laboratory corks and for transporting many kinds of liquids.
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.