Replaceable Junction Tool and Ceramic Junctions, METTLER TOLEDO®
Replaceable Ceramic Junctions
Catalog # CA11278-738
Supplier: Mettler Toledo
Fournisseur #: 51340268
Replaceable Junction Tool and Ceramic Junctions, METTLER TOLEDO®
Catalog # CA11278-738
Supplier: Mettler Toledo
Jump to:
Product Details & Documents
With a simple technique, this tool quickly replaces old, contaminated ceramic junctions from the InLab® 403 electrodes.
Spécifications
- Désignation:Replaceable Ceramic Junctions