Conductivity Standards, Sachets | Avantor
Conductivity Standards, Sachets
30×20 ml, 1413 uS/cm, Sachets
Catalog # CA11238-450
Supplier:   Mettler Toledo
Fournisseur #:  51302049
Conductivity Standards, Sachets
Catalog # CA11238-450
Supplier:  Mettler Toledo
CAS Number:  
Conductivity Standards, Sachets
Conductivity Standards, Sachets
30×20 ml, 1413 uS/cm, Sachets
CAS Number:  
Price
$169.00
Log in pour la disponibilité
Pack of 30
Product Details & Documents
NIST-traceable standard for conductivity electrode calibration and verification.
  • NIST-traceable
  • Ensure freshness
  • Improve data
Mettler Toledo sachets ensure a fresh portion of standard is used for each calibration, improving your conductivity and TDS data.
Spécifications
Spécifications
  • Type de conditionnement:
    Sachets
  • Taille:
    30×20 ml
  • Conductivity:
    1413 uS/cm