GLUCOSE DRINK SIMPLY 75G CS24
Catalog # CA11006-173
Supplier: Azer Scientific
GLUCOSE DRINK SIMPLY 75G CS24
Catalog # CA11006-173
Supplier: Azer Scientific
Supplier Number: 10-SP-075-CA
Frequently Bought Together
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
Peroxide Test, MQuant®, MilliporeSigma
Supplier: MilliporeSigma
Leakbuster™ Specimen Containers
Supplier: Starplex
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Cotton Tip Applicators, DUKAL™ Corporation
Supplier: Dukal
Acide chlorhydrique 6.0 N analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Contrad® 70 Liquid Detergent
Supplier: Decon Labs
Acétonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
GLUCOSE DRINK SIMPLY 50G CS24.
Supplier: Azer Scientific
Single-Use Needles, BD Medical
Supplier: BD
Bard-Parker® Protected Disposable Scalpels, Sterile, Aspen Surgical
Supplier: Aspen Surgical Products
VWR® 100-Hour Mini-Alarm Timer/Stopwatch
Supplier: VWR International
Eau for HPLC, Supelco®
Supplier: MilliporeSigma
Corning® Matrigel® Basement Membrane Matrix, Corning
Supplier: Corning