qScript™ cDNA Synthesis Kits, QuantaBio
qScript™ cDNA Synthesis Kits, QuantaBio
qScript™ cDNA Synthesis Kits, 100
Catalog # CA101414-098
Supplier:   Quantabio
Supplier #:  95047-100
qScript™ cDNA Synthesis Kits, QuantaBio
Catalog # CA101414-098
Supplier:  Quantabio
CAS Number:  
qScript™ cDNA Synthesis Kits, QuantaBio
qScript™ cDNA Synthesis Kits, QuantaBio
qScript™ cDNA Synthesis Kits, 100
CAS Number:  
Price
$556.06
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The qScript cDNA Synthesis Kit is a sensitive and easy-to-use solution for RNA quantification using two step RT-qPCR.
  • Economical 2-component kit ideally suited for high throughput expression studies
  • Optimized, double pre-primed master mix component ensures equal representation of 5′ and 3′ RNA sequences
  • Broad linear dynamic detection range with limiting (10pg) or plentiful (1ug) samples of total RNA
The qScript™ reaction mix provides all the necessary components for cDNA synthesis except the enzyme and RNA template. The optimized blend of random and oligo(dT) primers provides robust, consistent, and unbiased first-strand synthesis over a broad range of RNA template concentrations. qScript™ reverse transcriptase is a mixture of an engineered MMLV-RT and a ribonuclease inhibitor protein. The simplified reaction procedure is ideally suited for high throughput expression studies using RT-PCR. The resulting cDNA product is directly compatible with current real-time PCR methods or conventional end-point RT-PCR where the targets are ≤1kb in length.
Specifications
Specifications
  • Désignation:
    qScript™ cDNA Synthesis Kits
  • No. of Reactions:
    100