Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Stain Kits, BD Diagnostics

Stain Kits, BD Diagnostics

TB Fluorescent Stain Kit T, 250 ml (8.4 oz.)
Catalog # CA-90008-886
Supplier:   BD
Supplier #:  212521
Stain Kits, BD Diagnostics
Catalog # CA-90008-886
Supplier:  BD
CAS Number:  
Stain Kits, BD Diagnostics

TB Fluorescent Stain Kit T, 250 ml (8.4 oz.)

CAS Number:  
Price
$331.00
Log in pour la disponibilité
Each (250ml)
Product Details & Documents

These kits include all the necessary components for categorizing bacteria as gram-negative or gram-positive, a definitive characteristic in the organism identification process.

Fluorescent stain kit 90008-886 enables the observation of smears at a lower magnification, increasing the field of view and decreasing examination time. Kit includes one 250mL bottle each of the following: TB Auramine-Rhodamine T, TB Decolorizer TM, and TB Potassium Permanganate.

TB stain kit K 90008-888 includes one 250mL bottle each of TB Carbolfuschin KF, TB Decolorizer, and TB Brilliant Green K.

TB stain kit ZN 90008-884 includes one 250mL bottle each of TB Carbolfuschin ZN, TB Decolorizer, and TB Methylene Blue.

Ordering information: All stain kits come conveniently packaged in a compact tray and bottles feature no-drip “keep clean” cap spouts for dispensing.

Spécifications

Spécifications

  • Désignation:
    TB Fluorescent Stain Kit T
  • Taille:
    250 ml (8.4 oz.)

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.