Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
BBL™ Salt Broth, Modified, BD | Avantor

BBL™ Salt Broth, Modified, BD

BBL™ salt broth, modified, 100×2 ml, Size K
Catalog # CA-90004-756
Supplier:   BD
Fournisseur #:  297189
BBL™ Salt Broth, Modified, BD
Catalog # CA-90004-756
Supplier:  BD
CAS Number:  
BBL™ Salt Broth, Modified, BD

BBL™ salt broth, modified, 100×2 ml, Size K

CAS Number:  
Price
$750.00
Log in pour la disponibilité
Pack of 100 (2ml)
La quantité minimale de commande est de 1
Product Details & Documents

BBL™ Salt Broth (Modified) is a selective medium for the differentiation of Enterococcus species from the Streptococcus bovis group of streptococci.

  • Growth (turbidity) within 72 hours indicates that the strain is salt tolerant
  • Sometimes changes the indicator color from purple to yellow within 72 hours
  • Broth media containing 6.5% NaCl are used to differentiate enterococci by determining the salt tolerance

Digests of animal tissue provide nitrogen and carbon compounds essential for bacterial growth. Dextrose is an energy source.

Bromcresol purple is an indicator of acid production during bacterial metabolism. Sodium chloride serves as a selective and differential agent.

Spécifications

Spécifications

  • Désignation:
    BBL™ salt broth, modified
  • Taille:
    100×2 ml
  • Tube Size:
    Size K
  • Cat. No.:
    CA-90004-756
  • Storage conditions:
    2...25 °C

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.