BBL™ Saline, Normal, BD
BBL™ saline, normal, 100×5 ml, Size K
: BD
: 221819
Normal saline is used in procedures that require the use of an isotonic diluent.
- Ready to use
- Provide ensured cell integrity
- Used with a vast array of procedures
An isotonic diluent may be used for dilution of bacterial cells to provide a concentration suitable for microscopic observation, determination of cell numbers, analysis for genetic or metabolic properties, washing cells preparatory to study, or preparation of standardized inocula.
- Désignation:BBL™ saline, normal
- Taille:100×5 ml
- Tube Size:Size K