Spring Scales
8001-MA - 10 g (0.25 oz) graduation intervals, 250 g (9 oz.)
Catalog # 97027-720
Supplier:   Ohaus
 :  80000039
Spring Scales
Catalog # 97027-720
Supplier:  Ohaus
Spring Scales
8001-MA - 10 g (0.25 oz) graduation intervals, 250 g (9 oz.)
Price
$58.18
Log in for availability
Each
Product Details & Documents
Ideal for general weighing, force experiments, action-reaction demonstrations.
  • Dial plates display
  • Metric/ounce/newtons
Spring scale is made of ABS housing with metal face plate, pointer, spring and hook. Features large bright colored dial face, easy-to-read black graduations on a frosted aluminum background.
Specifications
  • Désignation:
    8001-MA - 10 g (0.25 oz) graduation intervals
  • Weighing Capacity:
    250 g (9 oz.)