Spring Scales
8001-MA - 10 g (0.25 oz) graduation intervals, 250 g (9 oz.)
Catalog # 97027-720
Supplier: Ohaus
: 80000039
Jump to:
Product Details & Documents
Ideal for general weighing, force experiments, action-reaction demonstrations.
- Dial plates display
- Metric/ounce/newtons
Spring scale is made of ABS housing with metal face plate, pointer, spring and hook. Features large bright colored dial face, easy-to-read black graduations on a frosted aluminum background.
Specifications
- Désignation:8001-MA - 10 g (0.25 oz) graduation intervals
- Weighing Capacity:250 g (9 oz.)